Learn More
Enzo Life Sciences 5(S),15(S)-DIHETE (50µg). CAS: 82200-87-1
SDP
Catalog No. 502010500
Encompass_Preferred
Biosynthesis in human eosinophils. Enhances neutrophil degranulation. Potent chemotaxin for human eosinophils. Formulation: Oil dissolved in ethanol. Purity: ≥99% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-0500 Supplier Enzo Life Sciences Supplier No. BMLD5150050
Biosynthesis in human eosinophils. Enhances neutrophil degranulation. Potent chemotaxin for human eosinophils. Formulation: Oil dissolved in ethanol. Purity: ≥99% (HPLC). Long Term Storage: -80°C.