Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-((S)-2,2-Dimethyltetrahydro-2H-pyran-4-yl)-1-((1S,2S)-2-methyl-1-(5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)cyclopropyl)-1H-indole-2-carboxylic acid | 2212021-83-3 | 99.96% | C22H25N3O5 | 250 MG
SDP

Catalog No. 5000358711
Encompass_Preferred

5-((S)-2,2-Dimethyltetrahydro-2H-pyran-4-yl)-1-((1S,2S)-2-methyl-1-(5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)cyclopropyl)-1H-indole-2-carboxylic acid (Compound 31k) is an intermediate of Orforglipron. This product is for research use only.

  • High purity at 99.96%
  • Available as a solid
  • White to off-white appearance
  • Store at room temperature for 3 years (solid)
  • Store at -80°C for 2 years or -20°C for 1 year (in solvent)

Catalog No. 50-003-58711 Supplier Medchemexpress LLC Supplier No. 250MGHY47663
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-((S)-2,2-Dimethyltetrahydro-2H-pyran-4-yl)-1-((1S,2S)-2-methyl-1-(5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)cyclopropyl)-1H-indole-2-carboxylic acid (Compound 31k) is an intermediate of Orforglipron. This product is for research use only.

  • High purity at 99.96%
  • Available as a solid
  • White to off-white appearance
  • Store at room temperature for 3 years (solid)
  • Store at -80°C for 2 years or -20°C for 1 year (in solvent)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.