Learn More
Enzo Life Sciences 5(S),6(R)-Dihydroxyeicosa-7E,9E,11Z,14Z-tetraenoic acid (DiHETE) (50µg). CAS: 82948-88-7
SDP
Supplier:  Enzo Life Sciences BMLDS0560050
Encompass_Preferred
Synthesis via enzymatic and nonenzymatic hydrolysis of LTA4. Induces contraction of guinea pig lung parenchymal strips. Partial agonist at LTD4 receptors. Potent inhibitor of LTB4 induced neutrophil migration (IC50 = 8nM). Alternative name: 5(S),6(R)-DiHETE. Formulation: Oil dissolved in ethanol. Purity: ≥99% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-0519