Learn More
Cayman Chemical 5(S),6(R)-7-trihydroxymethyl Heptanoate, Purity: ≥95%, CAS: 78606-80-1, 5 mg
SDP
Catalog No. 502891165
Encompass_Preferred
A potent bioactive analog of lipoxin A4; inhibits LTB4-induced chemotaxis of PMNs (IC50 = 5 nM)
Catalog No. 50-289-1165 Supplier Cayman Chemical Supplier No. 100050325 MG
A potent bioactive analog of lipoxin A4; inhibits LTB4-induced chemotaxis of PMNs (IC50 = 5 nM)