Learn More
Medchemexpress LLC 5(S)-HETE-d8 | 330796-62-8 | 96.1% | 328.52 | 25 UG
SDP
Catalog No. 5000439649
Encompass_Preferred
5(S)-HETE-d8 is a deuterium-labeled form of 5(S)-HETE, designed for research purposes. It can serve as a tracer and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for quantitation during drug development, with the potential to influence pharmacokinetic and metabolic profiles of drugs.
  • Research use only
  • Tracer
  • Internal standard
  • Deuterium labeled
Catalog No. 50-004-39649 Supplier Medchemexpress LLC Supplier No. 25UGHY113434S
5(S)-HETE-d8 is a deuterium-labeled form of 5(S)-HETE, designed for research purposes. It can serve as a tracer and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for quantitation during drug development, with the potential to influence pharmacokinetic and metabolic profiles of drugs.
  • Research use only
  • Tracer
  • Internal standard
  • Deuterium labeled