Learn More
Seracare Life Sciences Inc 5% SDS
Catalog No. 5067490
Encompass_Preferred
5% SDS is a detergent surfactant commonly used in preparing proteins for sodium dodecyl sulfate – polyacrylamide gel electrophoresis (SDS-PAGE) of proteins and as an additive to transfer buffer in Western blotting. SDS increases the elution rate of proteins from the gel.
This item is not returnable. View return policy
Catalog No. 50-674-90 Supplier Seracare Life Sciences Inc Supplier No. 54600046
5% SDS is a detergent surfactant commonly used in preparing proteins for sodium dodecyl sulfate – polyacrylamide gel electrophoresis (SDS-PAGE) of proteins and as an additive to transfer buffer in Western blotting. SDS increases the elution rate of proteins from the gel.