Learn More
AdipoGen 5-SFX 5-FAM-X
SDP
Supplier:  AdipoGen CDXF0036M050
Encompass_Preferred
Chemical. CAS 194661-60-4. Formula C27H23NO8. MW 489.47. Synthetic. Used as a fluorescein-based fluorescent label for proteins that can be analyzed in applications such as quenching experiments. Spectral data lambdaex 491nm lambdaem 515nm 0.1 M Tris, pH 9.0.
Catalog No. 50-256-0922