Learn More
Medchemexpress LLC 5-Sulfosalicylic acid dihydrate | 5965-83-3 | >99.88% | 1000 G
SDP
Catalog No. 5000409473
Encompass_Preferred
5-Sulfosalicylic acid dihydrate is a sulfonated salicylic acid derivative effective against the breast cancer cell line MCF-7 with less toxicity. It also exhibits antioxidant activities and is supplied as a white to off-white solid.
  • Effective against breast cancer cell line MCF-7
  • Exhibits antioxidant activities
  • Supplied as a white to off-white solid
  • Molecular weight: 254.21
  • For research use only
Catalog No. 50-004-09473 Supplier Medchemexpress LLC Supplier No. HYB08121000G
5-Sulfosalicylic acid dihydrate is a sulfonated salicylic acid derivative effective against the breast cancer cell line MCF-7 with less toxicity. It also exhibits antioxidant activities and is supplied as a white to off-white solid.
  • Effective against breast cancer cell line MCF-7
  • Exhibits antioxidant activities
  • Supplied as a white to off-white solid
  • Molecular weight: 254.21
  • For research use only