Learn More
Medchemexpress LLC 5-Sulfosalicylic acid dihydrate | 5965-83-3 | 99.9% | 10 MM 1 ML
SDP
Catalog No. 5000408095
Encompass_Preferred
5-Sulfosalicylic acid dihydrate is a sulfonated salicylic acid derivative. It is effective against the breast cancer cell line, MCF-7, with less toxicity and possesses antioxidant activities.
  • It is a sulfonated salicylic acid derivative.
  • It demonstrates effectiveness against the breast cancer cell line, MCF-7, with reduced toxicity.
  • It possesses antioxidant activities.
  • It can reduce the viability of MCF-7 cells in vitro.
  • It is for research use only.
Catalog No. 50-004-08095 Supplier Medchemexpress LLC Supplier No. HYB081210MM/1ML
5-Sulfosalicylic acid dihydrate is a sulfonated salicylic acid derivative. It is effective against the breast cancer cell line, MCF-7, with less toxicity and possesses antioxidant activities.
  • It is a sulfonated salicylic acid derivative.
  • It demonstrates effectiveness against the breast cancer cell line, MCF-7, with reduced toxicity.
  • It possesses antioxidant activities.
  • It can reduce the viability of MCF-7 cells in vitro.
  • It is for research use only.