Learn More
Medchemexpress LLC 5-Sulfosalicylic acid (dihydrate) | 5965-83-3 | 99.9% | 50 G
SDP
Catalog No. 5000409296
Encompass_Preferred
5-Sulfosalicylic acid dihydrate is a sulfonated salicylic acid derivative that demonstrates effectiveness against the breast cancer cell line MCF-7 with reduced toxicity and possesses antioxidant activities.
  • Effective against breast cancer cell line MCF-7
  • Reduced toxicity
  • Possesses antioxidant activities
  • Reduces cell viability in MCF-7 cells at 0.5-4 mM concentrations
  • Leads to 63.3% viability in MCF-7 cells at 1 mM and 70.4% in HUVEC control cells after 24 hours
Catalog No. 50-004-09296 Supplier Medchemexpress LLC Supplier No. HYB081250G
5-Sulfosalicylic acid dihydrate is a sulfonated salicylic acid derivative that demonstrates effectiveness against the breast cancer cell line MCF-7 with reduced toxicity and possesses antioxidant activities.
  • Effective against breast cancer cell line MCF-7
  • Reduced toxicity
  • Possesses antioxidant activities
  • Reduces cell viability in MCF-7 cells at 0.5-4 mM concentrations
  • Leads to 63.3% viability in MCF-7 cells at 1 mM and 70.4% in HUVEC control cells after 24 hours