Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Sulfosalicylic acid dihydrate, ACS, 99+%

Catalog No. aa4314414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4314414 25 g
AA4314422 100 g
AA4314436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4314414 Supplier Thermo Scientific Chemicals Supplier No. 04314414
Only null left
Add to Cart

Reagent for precipitation of proteins | CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol

Reagent for precipitation of proteins5-Sulfosalicylic acid dihydrate is used as a metal scavenger and it forms proton-transfer dye complexes with diazo compounds such as 4-(phenyldiazenyl)aniline. It is employed as a preparative measure for removing proteins prior to chromatographic analysis. It plays a vital role in reducing and fixation of proteins in agarose and polyacrylamide gels. It acts as a surfactant and lubricating grease additive. It is also used in urine tests to determine urine protein content. Further, it acts as an intermediate in the synthesis of active pharmaceutical ingredients like doxycycline and methacycline. In addition to this, it is used as a spray reagent for the detection of amino acids on thin layer chromatography (TLC) plates and as a doping agent for polyaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5965-83-3
Molecular Formula C9H14O6S
Molecular Weight (g/mol) 250.27
MDL Number MFCD00007508,MFCD00149540
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
Synonym 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
PubChem CID 2723734
IUPAC Name 2-hydroxy-5-sulfobenzoic acid;dihydrate
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O

Specifications

Melting Point 103°C to 112°C
Quantity 25 g
UN Number UN2585
Beilstein 650741
Sensitivity Light sensitive
Merck Index 14,8964
Solubility Information Soluble in water,alcohol,ether and polar solvents.
Formula Weight 254.22 (218.19 Anhydrous)
Percent Purity ≥99%
Grade ACS
Physical Form Powder
Chemical Name or Material 5-Sulfosalicylic Acid, Dihydrate
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
EINECSNumber 202-555-6
RTECSNumber VO6500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.