Learn More
Biotium 5-CARBOXYTetramethylrhodamine, SINGLE IS
Catalog No. 501964800
Encompass_Preferred
Free acid form of 5-TAMRA (5-Carboxytetramethylrhodamine, single isomer). Properties: Ex/ Em (MeOH) = 540/565 nm; Dark red solid soluble in DMSO, DMF, or MeOH C 25 H 22 N 2 O 5; MW: 430.5; [91809-66-4]
Catalog No. 50-196-4800 Supplier Biotium Supplier No. 90009
Add to Cart
Edge
Add to Cart
Free acid form of 5-TAMRA (5-Carboxytetramethylrhodamine, single isomer). Properties: Ex/ Em (MeOH) = 540/565 nm; Dark red solid soluble in DMSO, DMF, or MeOH C 25 H 22 N 2 O 5; MW: 430.5; [91809-66-4]