Learn More
Abcam 5-TAMRA (5-Carbo x ytetramethylrhodamine), Fluorescent dye for DNA, peptides and proteins, 25MG
Catalog No. 502705192
Encompass_Preferred
MW 430.45 g/mol, Purity >97%. Fluorescent dye for DNA, peptides and proteins. Amine-reactive. Used for oligonucleotide labeling and automated DNA sequencing applications. TAMRA is often used as FRET acceptor for FAM fluorophore. Free 5(6)-TAMRA (ab145282) and 6-TAMRA (ab145338) isomer. 5-TAMRA N-succinimidyl ester (ab145433) precursor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5192 Supplier Abcam Supplier No. AB14543825MG
MW 430.45 g/mol, Purity >97%. Fluorescent dye for DNA, peptides and proteins. Amine-reactive. Used for oligonucleotide labeling and automated DNA sequencing applications. TAMRA is often used as FRET acceptor for FAM fluorophore. Free 5(6)-TAMRA (ab145282) and 6-TAMRA (ab145338) isomer. 5-TAMRA N-succinimidyl ester (ab145433) precursor.
The product is subject to the following: Abcam Restricted Use Statement