Learn More
AdipoGen 5-TAMRA
SDP
Catalog No. 502561507
Encompass_Preferred
Chemical. CAS 91809-66-4. Formula C25H22N2O5. MW 430.45. Synthetic. Free acid form of 5-carboxytetramethylrhodamine single isomer. Widely used fluorophore for preparing bioconjugates, especially fluorescent antibody and avidin derivatives used in immunochemistry. Used as a dye for oligonucleotide labeling and automated DNA sequencing applications.
Catalog No. 50-256-1507 Supplier AdipoGen Supplier No. CDXC0055M025
Chemical. CAS 91809-66-4. Formula C25H22N2O5. MW 430.45. Synthetic. Free acid form of 5-carboxytetramethylrhodamine single isomer. Widely used fluorophore for preparing bioconjugates, especially fluorescent antibody and avidin derivatives used in immunochemistry. Used as a dye for oligonucleotide labeling and automated DNA sequencing applications.