Learn More
AdipoGen 5-TAMRA Maleimide
SDP
Supplier:  AdipoGen CDXT0029M025
Encompass_Preferred
Chemical. CAS 174568-67-3. Formula C28H23N3O5. MW 481.5. Synthetic. Labeling reagent for protein thiols for cytofluorescence.
Catalog No. 50-256-0043