Learn More
AdipoGen 5-TAMRA N-succinimidyl ester
SDP
Catalog No. 502561503
Encompass_Preferred
Chemical. CAS 150810-68-7. Formula C29H25N3O7. MW 527.52. Synthetic. Amine-reactive form of 5-carboxytetramethylrhodamine single isomer. Conjugates yield bright, pH-insensitive orange-red fluorescence Ex/Em ~546/579nm with good photostability.
Catalog No. 50-256-1503 Supplier AdipoGen Supplier No. CDXC0057M001
Chemical. CAS 150810-68-7. Formula C29H25N3O7. MW 527.52. Synthetic. Amine-reactive form of 5-carboxytetramethylrhodamine single isomer. Conjugates yield bright, pH-insensitive orange-red fluorescence Ex/Em ~546/579nm with good photostability.