Learn More
Abcam 5-TAMRA N-succinimidyl ester (5-Carbo x y-tetramethylrhodamine N-succinimidyl ester), 1MG
Catalog No. 502705196
Encompass_Preferred
MW 527.52 g/mol, Purity >97%. Amine-reactive fluorescent probe. 5-TAMRA (ab145438) derivative. 6-TAMRA N-succinimidyl ester (ab145334) isomer. Suitable for applications as an enzyme substrate, molecular probe, molecular beacons and FRET partner.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5196 Supplier Abcam Supplier No. AB1454331MG
MW 527.52 g/mol, Purity >97%. Amine-reactive fluorescent probe. 5-TAMRA (ab145438) derivative. 6-TAMRA N-succinimidyl ester (ab145334) isomer. Suitable for applications as an enzyme substrate, molecular probe, molecular beacons and FRET partner.
The product is subject to the following: Abcam Restricted Use Statement