Learn More
Biotium 5-TAMRA-PEO3-amine
Catalog No. 501964824
Encompass_Preferred
5-TAMRA-PEO3-amine (Tetramethylrhodamine-PEO3-amine) has a long water-soluble PEG spacer separating the dye and the amine functional group. The dye can be coupled to activated carboxy groups or sulfonyl chlorides. Properties: Ex/Em (MeOH) = 544/571 nm; Dark purple solid soluble in DMSO, DMF, or H2O; Store at 4°C and protect from light; C 37 H 42 F 6 N 4 O 11; MW: 832.74
Catalog No. 50-196-4824 Supplier Biotium Supplier No. 90107
5-TAMRA-PEO3-amine (Tetramethylrhodamine-PEO3-amine) has a long water-soluble PEG spacer separating the dye and the amine functional group. The dye can be coupled to activated carboxy groups or sulfonyl chlorides. Properties: Ex/Em (MeOH) = 544/571 nm; Dark purple solid soluble in DMSO, DMF, or H2O; Store at 4°C and protect from light; C 37 H 42 F 6 N 4 O 11; MW: 832.74