Learn More
Aat Bioquest 5-TAMRA SE [5-Carboxytetramethylrhodamine succinimidyl ester] CAS 150810-68-7 (100 mg)
SDP
Catalog No. 502249339
Encompass_Preferred
The single TAMRA isomers are increasingly preferred for labeling peptides and nucleotides because they give better resolution in HPLC purification that is often required in the conjugation processes. This catalog is for 100 mg.
This item is not returnable. View return policy
Catalog No. 50-224-9339 Supplier Aat Bioquest Supplier No. 374
The single TAMRA isomers are increasingly preferred for labeling peptides and nucleotides because they give better resolution in HPLC purification that is often required in the conjugation processes. This catalog is for 100 mg.