Learn More
Invitrogen™ 5-TAMRA, SE (5-Carboxytetramethylrhodamine, Succinimidyl Ester), single isomer
Description
The amine-reactive 5-TAMRA, SE and its conjugates yield bright, pH-insensitive orange-red fluorescence (approximate excitation/emission maxima ∼546/579) with good photostability.
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | 5-TAMRA SE |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Active Ester, Succinimidyl Ester |
| Label or Dye | TAMRA™ Isomers, TMR (Tetramethylrhodamine) |
| Label Type | Classic Dyes |
| Emission | 579 |
| Excitation | 546 |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.