Learn More
Aat Bioquest 5-TAMRA, SE [5-Carboxytetramethylrhodamine, succinimidyl ester] *CAS#: 150810-68-7*
SDP
Catalog No. 501952861
Encompass_Preferred
The single TAMRA isomers are increasingly preferred for labeling peptides and nucleotides because they give better resolution in HPLC purification that is often required in the conjugation processes.
Catalog No. 50-195-2861 Supplier Aat Bioquest Supplier No. 373
The single TAMRA isomers are increasingly preferred for labeling peptides and nucleotides because they give better resolution in HPLC purification that is often required in the conjugation processes.