Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-tert-Butyl-2-methylbenzenethiol 90.0+%, TCI America™
SDP

Catalog No. B169125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B169125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B169125ML Supplier TCI America Supplier No. B169125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7340-90-1
Molecular Formula C11H16S
Molecular Weight (g/mol) 180.309
MDL Number MFCD00142942
InChI Key ZSBNXOIAJFVXMP-UHFFFAOYSA-N
Synonym 5-tert-butyl-2-methylthiophenol, 4-tert-butyl-2-mercaptotoluene, 5-tert-butyl-o-thiocresol, benzenethiol, 5-1,1-dimethylethyl-2-methyl, acmc-1bejs, zsbnxoiajfvxmp-uhfffaoysa, 2-methyl-5-tert-butyl thiophenol, 2-methyl-5-tert-butylbenzenethiol, 2-methyl-5-tert.-butyl-thiophenol, 5-tert-butyl-2-methylbenzenethiol
PubChem CID 596868
IUPAC Name 5-tert-butyl-2-methylbenzenethiol
SMILES CC1=C(C=C(C=C1)C(C)(C)C)S

Specifications

Color Yellow
Quantity 25 mL
UN Number 3082
Formula Weight 180.31
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-tert-Butyl-2-methylbenzenethiol
EINECSNumber (3)-2896
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.