Learn More
Medchemexpress LLC 5-(tert-Butyl)isobenzofuran-1,3-dione | 32703-79-0 | 96.0% | 204.23 | 5 G
SDP
Catalog No. 5000453070
Encompass_Preferred
5-(tert-Butyl)isobenzofuran-1,3-dione is an intermediate and dye, also known as 4-tert-Butylphthalic Anhydride. It is intended for research use only and not for medical applications.
  • Intermediate and dye
  • For research use only
  • Fluorescent dye target
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years, or at -20°C for 1 year
Catalog No. 50-004-53070 Supplier Medchemexpress LLC Supplier No. HYW0124825G
5-(tert-Butyl)isobenzofuran-1,3-dione is an intermediate and dye, also known as 4-tert-Butylphthalic Anhydride. It is intended for research use only and not for medical applications.
  • Intermediate and dye
  • For research use only
  • Fluorescent dye target
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years, or at -20°C for 1 year