Learn More
Medchemexpress LLC 5-thiazoleethanol, 4-methyl | 137-00-8 | MFCD00005339 | 99.5% | 143.21 g/mol | C6H9NOS | 50 G
SDP
Catalog No. 5000300671
Encompass_Preferred
4-Methyl-5-thiazoleethanol is a sulfur-containing thiazole alcohol used as a flavor compound and analytical reference. It has a beef- and nut-like aroma and is supplied for research use with high stated purity.
  • Natural sulfur-containing flavor compound with beef- and nut-like aroma.
  • High purity (99.53%) suitable for research and analytical use.
  • Molecular formula C6H9NOS; molecular weight 143.21 g/mol.
  • Available in solid pack sizes and as DMSO solution for analytical preparation.
  • Commonly used as a reference standard and in flavor chemistry studies.
Catalog No. 50-003-00671 Supplier Medchemexpress LLC Supplier No. HYW01569550G
Add to Cart
Add to Cart
4-Methyl-5-thiazoleethanol is a sulfur-containing thiazole alcohol used as a flavor compound and analytical reference. It has a beef- and nut-like aroma and is supplied for research use with high stated purity.
  • Natural sulfur-containing flavor compound with beef- and nut-like aroma.
  • High purity (99.53%) suitable for research and analytical use.
  • Molecular formula C6H9NOS; molecular weight 143.21 g/mol.
  • Available in solid pack sizes and as DMSO solution for analytical preparation.
  • Commonly used as a reference standard and in flavor chemistry studies.