Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(thien-2-yl)thiophene-2-carbonitrile, Thermo Scientific™

Catalog No. SEW02204DE
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SEW02204DE 5 g
SEW02204CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SEW02204DE Supplier Thermo Scientific Chemicals Supplier No. SEW02204DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16278-99-2
Molecular Formula C9H5NS2
Molecular Weight (g/mol) 191.266
InChI Key ANARYBGZNUMARH-UHFFFAOYSA-N
Synonym 5-thien-2-yl thiophene-2-carbonitrile, 2,2'-bithiophene-5-carbonitrile, 5-thiophen-2-yl thiophene-2-carbonitrile, maybridge3_006613, 5-cyano-2,2'-bithiophene, 5-2-thienyl thiophene-2-carbonitrile
PubChem CID 2739839
IUPAC Name 5-thiophen-2-ylthiophene-2-carbonitrile
SMILES C1=CSC(=C1)C2=CC=C(S2)C#N

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material 5-(Thien-2-yl)thiophene-2-carbonitrile
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.