Learn More
LGC GENOMICS LLC 5'-Thiol Modifier C6 S-S CE-Phosphoramidite, Thiol-Reactive Group, Robust Conjugation Method, 10.74074074074074 g
Catalog No. NC3443399
Encompass
The 5'-Thiol Modifier C6 S-S CE-Phosphoramidite is a versatile chemical reagent designed for the targeted incorporation of thiol groups into oligonucleotides, which allows for easy post-synthesis modifications. Its unique disulfide bridge and C6 spacer facilitate effective thiol liberation, enhancing conjugation efficiency for various applications including biosensing and microarrays. Unlike other modifiers, it simplifies purification processes, making it suitable for diverse bioconjugation needs.
Catalog No. NC3443399 Supplier LGC GENOMICS LLC Supplier No. LK2126C001
The 5'-Thiol Modifier C6 S-S CE-Phosphoramidite is a versatile chemical reagent designed for the targeted incorporation of thiol groups into oligonucleotides, which allows for easy post-synthesis modifications. Its unique disulfide bridge and C6 spacer facilitate effective thiol liberation, enhancing conjugation efficiency for various applications including biosensing and microarrays. Unlike other modifiers, it simplifies purification processes, making it suitable for diverse bioconjugation needs.