Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Trifluoromethoxy)-1H-indole-2-carboxylic acid, 97%, Thermo Scientific™

Catalog No. KM09272CB
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KM09272CB 250 mg
KM09272DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. KM09272CB Supplier Thermo Scientific Chemicals Supplier No. KM09272CB
Add to Cart

Chemical Identifiers

CAS 175203-84-6
Molecular Formula C10H6F3NO3
Molecular Weight (g/mol) 245.16
MDL Number MFCD00276997
InChI Key DJJJSHFPEISHFN-UHFFFAOYSA-N
Synonym 5-trifluoromethoxy-1h-indole-2-carboxylic acid, 5-trifluoromethoxy indole-2-carboxylic acid, maybridge1_004876, 1h-indole-2-carboxylic acid, 5-trifluoromethoxy, 5-trifluoromethyloxy-1h-indole-2-carboxylic acid
PubChem CID 2777290
IUPAC Name 5-(trifluoromethoxy)-1H-indole-2-carboxylic acid
SMILES OC(=O)C1=CC2=CC(OC(F)(F)F)=CC=C2N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 245.16
Percent Purity 97%
Chemical Name or Material 5-(Trifluoromethoxy)-1H-indole-2-carboxylic acid
missing translation for 'healthHazard2' GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
missing translation for 'healthHazard3' GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.