Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Trifluoromethoxy)salicylaldehyde, 98+%, Thermo Scientific Chemicals

Catalog No. AAL1713603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1713603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1713603 Supplier Thermo Scientific Chemicals Supplier No. L1713603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 93249-62-8 | Molecular Formula: C8H5F3O3 | Molecular Weight: 206.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93249-62-8
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD00075249
InChI Key WQUZBERVMUEJTD-UHFFFAOYSA-N
Synonym 2-hydroxy-5-trifluoromethoxy benzaldehyde, 5-trifluoromethoxy salicylaldehyde, benzaldehyde, 2-hydroxy-5-trifluoromethoxy, acmc-20antc, 5-trifluoromethoxysalicylaldehyde, 5-trifluoromethoxy-salicylaldehyde, 2-hydroxy-5-trifluoromethoxy-benzaldehyde, 20hydroxy-5-trifluoromethoxy benzaldehyde, 2-oxidanyl-5-trifluoromethyloxy benzaldehyde, benzaldehyde,2-hydroxy-5-trifluoromethoxy
PubChem CID 1268058
IUPAC Name 2-hydroxy-5-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=C(C=C1OC(F)(F)F)C=O)O

Specifications

Melting Point 31°C to 33°C
Boiling Point 82°C (60 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 7023825
Sensitivity Air Sensitive
Formula Weight 206.12
Percent Purity ≥98%
Chemical Name or Material 5-(Trifluoromethoxy)salicylaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.