Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-(Trifluoromethyl)benzo[d]oxazol-2(3H)-one

Catalog No. JRD03701G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD03701G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD03701G Supplier Sigma Aldrich Supplier No. JRD03701G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-ethynylthiophene may be used in the synthesis of the following:
  • 1-(2-bromobenzyl)-4-(thiophen-3-yl)-1h-1,2,3-triazole which is obtained by heating with 2-iodophenylethylazide in the presence of copper acetate monohydrate catalyst in n-methyl-2-pyrrolidone
  • N-benzyl-1-phenyl-5-(thiophen-3-yl)-4-pentyn-2-amine via a multi-step reaction process
  • [(c4h3s-3)cgdfcag]n, a polymeric compound obtained via reaction with silver nitrate in the presence of triethylamine in acetonitrile
  • 4,5-bis(thiophen-3-ylethynyl)phthalonitrile via sonogashira cross-coupling reaction with 4,5-dichlorophthalonitrile

Specifications

CAS 14733-68-7
Molecular Formula C8H4F3NO2
Linear Formula C8H4F3NO2
MDL Number MFCD01664311
Quantity 5 g
Molecular Weight (g/mol) 203.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.