Learn More
Abcam 5-TRITC (Tetramethylrhodamine-5-isothiocyanate), Amine and nucleic acid reactive fluorescent probe, 25MG
Catalog No. 502705171
Encompass_Preferred
MW 443.52 g/mol, Purity >95%. Amine and nucleic acid reactive fluorescent probe. 6-TRITC (ab145403) and 5(6)-TRITC (ab145353) S-isomer. Spectrally similar to TAMRA (ab145282, ab145438, ab145338, ab146486, ab145334, ab145433). Biodistributes covalently linked to poly(lactic-co-glycolic) acid (PLGA) and PLGA/chitosan nanoparticles after oral administration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5171 Supplier Abcam Supplier No. AB14547225MG
Add to Cart
Add to Cart
MW 443.52 g/mol, Purity >95%. Amine and nucleic acid reactive fluorescent probe. 6-TRITC (ab145403) and 5(6)-TRITC (ab145353) S-isomer. Spectrally similar to TAMRA (ab145282, ab145438, ab145338, ab146486, ab145334, ab145433). Biodistributes covalently linked to poly(lactic-co-glycolic) acid (PLGA) and PLGA/chitosan nanoparticles after oral administration.
The product is subject to the following: Abcam Restricted Use Statement