Learn More
Cayman Chemical 5-trans Fluprostenol, Purity: ≥95%, CAS: 57968-83-9, 10 mg
SDP
Catalog No. 502966771
Encompass_Preferred
A potential impurity in commercial preparations of fluprostenol
Catalog No. 50-296-6771 Supplier Cayman Chemical Supplier No. 1677610 MG
A potential impurity in commercial preparations of fluprostenol