Learn More
Cayman Chemical 5-trns Latnoprost 10mg
SDP
Supplier:  Cayman Chemical 1681310 MG
Encompass_Preferred
A derivative of latanoprost (free acid), an isomer of latanoprost, and an analytical standard; may have similar biological activities as F-type prostaglandin cis isomers; a potential impurity in commercial preparations of latanoprost; intended as an analytical standard
Catalog No. 50-296-4911