Learn More
Cayman Chemical 5-trans Latanoprost, Purity: ≥98%, CAS: 913258-34-1, 50 mg
SDP
Catalog No. 502966563
Encompass_Preferred
A derivative of latanoprost (free acid), an isomer of latanoprost, and an analytical standard; may have similar biological activities as F-type prostaglandin cis isomers; a potential impurity in commercial preparations of latanoprost; intended as an analytical standard
Catalog No. 50-296-6563 Supplier Cayman Chemical Supplier No. 1681350 MG
A derivative of latanoprost (free acid), an isomer of latanoprost, and an analytical standard; may have similar biological activities as F-type prostaglandin cis isomers; a potential impurity in commercial preparations of latanoprost; intended as an analytical standard