Learn More
Cayman Chemical 5-trns U-44069 10mg
SDP
Catalog No. 502970945
Encompass_Preferred
The trans isomer of U-44069; inhibits mPGES at 10 µM
Catalog No. 50-297-0945 Supplier Cayman Chemical Supplier No. 1644210 MG
The trans isomer of U-44069; inhibits mPGES at 10 µM