Learn More
Medchemexpress LLC 5-Ureidohydantoin (Standard) | 97-59-6 | MFCD00005260 | 158.12 | 50 MG
SDP
Catalog No. 5000388803
Encompass_Preferred
Allantoin (Standard) is the analytical standard of Allantoin. It is a skin conditioning agent that promotes healthy skin, stimulates new and healthy tissue growth. This product is intended for research and analytical applications. The compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard for research and analytical applications.
  • Promotes healthy skin.
  • Stimulates new and healthy tissue growth.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-88803 Supplier Medchemexpress LLC Supplier No. HYN0543R50MG
Allantoin (Standard) is the analytical standard of Allantoin. It is a skin conditioning agent that promotes healthy skin, stimulates new and healthy tissue growth. This product is intended for research and analytical applications. The compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard for research and analytical applications.
  • Promotes healthy skin.
  • Stimulates new and healthy tissue growth.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.