Learn More
Starna Cells Inc 5 WINDOW QTZ CUVETTE GRD SEAL
SDP
Catalog No. NC9265851
Encompass_Preferred
Fluorimeter, Macro/Standard Rectangular, Spectrosil quartz, 10 W Internal, 12.5L x 12.5W x 45H, 3.500mL, Open top, with non-sealing PTFE cover, Four windows and base polished
Catalog No. NC9265851 Supplier Starna Cells Inc Supplier No. 3Q10/GS
Fluorimeter, Macro/Standard Rectangular, Spectrosil quartz, 10 W Internal, 12.5L x 12.5W x 45H, 3.500mL, Open top, with non-sealing PTFE cover, Four windows and base polished