Learn More
Polysciences, Inc. Glutaraldehyde, 25% EM Grade (Glass Distilled), 111-30-8, MFCD00007025, 5 X 100 M
SDP
Catalog No. NC9072609
Encompass
Glutaraldehyde, Molecular Formula: C5 H8 O2, Molecular Weight: 100.116, Refractive Index: 1.375, Density: 1.062. Hazards: Irritant , Skin sensitizer , Toxic (Ingestion), Handling: Glove, chemical goggles & fume hood, Storage: Store at 4 degrees celsius. Polysciences was the first to develop a superior grade of glutaraldehyde suitable for immunochemical techniques. Each batch of vacuum distilled glutaraldehyde manufactured by Polysciences is analyzed to maintain consistent high quality and freedom from contaminants which destroy antigenicity. Each ampoule is fitted with an ampoule cracker for added safety. EM grade is recommended for histochemical or immunological techniques.
Catalog No. NC9072609 Supplier Polysciences, Inc. Supplier No. 019095
Glutaraldehyde, Molecular Formula: C5 H8 O2, Molecular Weight: 100.116, Refractive Index: 1.375, Density: 1.062. Hazards: Irritant , Skin sensitizer , Toxic (Ingestion), Handling: Glove, chemical goggles & fume hood, Storage: Store at 4 degrees celsius. Polysciences was the first to develop a superior grade of glutaraldehyde suitable for immunochemical techniques. Each batch of vacuum distilled glutaraldehyde manufactured by Polysciences is analyzed to maintain consistent high quality and freedom from contaminants which destroy antigenicity. Each ampoule is fitted with an ampoule cracker for added safety. EM grade is recommended for histochemical or immunological techniques.