Learn More
Alkali Scientific 5 X 7 inch BioBlue-MR™ Autoradiography Film, 100 Sheets
SDP
Catalog No. 502335868
Encompass_Preferred
BioBlue-MR is a high quality, blue sensitive film that is ideal for a wide range of electrophoretic and autoradiography applications. It can be used with calcium tungstate or blue-emitting rare-earth screens. BioBlue-MR exhibits excellent sensitivity and contrast characteristics and is coated on both sides for improved consistency and overall performance. It reliably produces sharp, bright images that are easily visualized and suitable for publication. Especially useful with chemiluminescence, BioBlue-MR autoradiography film works well with 14C, 32P, 33P, 125I, and 35S. Compatible with automatic processors and manual development methods, Bioblue-XR compares directly with films costing up to 3 times as much. With a shelf life of 2 years from date of production, BioBlue-XR is packaged in lined, light-safe dispenser boxes of 100 sheets per box.
Catalog No. 50-233-5868 Supplier Alkali Scientific Supplier No. XR1570
BioBlue-MR is a high quality, blue sensitive film that is ideal for a wide range of electrophoretic and autoradiography applications. It can be used with calcium tungstate or blue-emitting rare-earth screens. BioBlue-MR exhibits excellent sensitivity and contrast characteristics and is coated on both sides for improved consistency and overall performance. It reliably produces sharp, bright images that are easily visualized and suitable for publication. Especially useful with chemiluminescence, BioBlue-MR autoradiography film works well with 14C, 32P, 33P, 125I, and 35S. Compatible with automatic processors and manual development methods, Bioblue-XR compares directly with films costing up to 3 times as much. With a shelf life of 2 years from date of production, BioBlue-XR is packaged in lined, light-safe dispenser boxes of 100 sheets per box.