Learn More
Chemglass Life Sciences Graded Seal, Quartz to Borosilicate, 5mm ID X 7mm OD
SDP
Catalog No. CGQ031004
Encompass_Preferred
Graded Seal, Quartz to Borosilicate, 5mm ID X 7mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.
Catalog No. CGQ031004 Supplier Chemglass Life Sciences Supplier No. CGQ031004
Graded Seal, Quartz to Borosilicate, 5mm ID X 7mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.