Learn More
Cayman Chemical 5 Z 8 Z 11 ZEIcosatrIenoI 10mg
SDP
Supplier:  Cayman Chemical 9019510 MG
Encompass_Preferred
A prolyl endopeptidase inhibitor (Kis 0.35 and 0.5 nM for the mouse and human enzymes respectively) inhibits the formation of angiotensin (1-7) from angiotensin II in kidney sections isolated from wild-type and Ace2-/- mice at 4 and 40 UM reduces hippocampal neuronal cell death in a rat model of global ischemia at 10 and 50 mg/kg
Catalog No. 50-289-5462