Learn More
Cayman Chemical 5(Z),8(Z),11(Z)-Eicosatrienoic Acid methyl ester, Purity: ≥98%, CAS: 14602-39-2, 10 mg
SDP
Catalog No. 502895291
Encompass_Preferred
Used as a standard for the analysis of fatty acids, when the fatty acids have been transesterified to methyl esters before analysis
Catalog No. 50-289-5291 Supplier Cayman Chemical Supplier No. 900021510 MG
Used as a standard for the analysis of fatty acids, when the fatty acids have been transesterified to methyl esters before analysis