Learn More
Cayman Chemical 5 Z 8 Z 11 ZEIcosatrIenoI 10mg
SDP
Catalog No. 502895064
Encompass_Preferred
Eicosatrienoic acid accumulates in the tissues of animals fed diets deficient in both ω-3 and ω-6 fatty acids. It can be converted to 3-series cysteinyl-leukotrienes but cannot serve as a COX substrate.{4349} The kidneys from essential fatty acid deficient rats are less immunogenic when transplanted, and the metabolites of eicosatrienoic acid are believed to play a role in altering the immune status of these organs.{712}
Catalog No. 50-289-5064 Supplier Cayman Chemical Supplier No. 9019010 MG
Eicosatrienoic acid accumulates in the tissues of animals fed diets deficient in both ω-3 and ω-6 fatty acids. It can be converted to 3-series cysteinyl-leukotrienes but cannot serve as a COX substrate.{4349} The kidneys from essential fatty acid deficient rats are less immunogenic when transplanted, and the metabolites of eicosatrienoic acid are believed to play a role in altering the immune status of these organs.{712}