Learn More
Cayman Chemical 5 Z 8 Z 11 ZEicosatrienoic Ac
SDP
Catalog No. 502887197
Encompass_Preferred
An internal standard for the quantification of 5(Z),8(Z),11(Z)-eicosatrienoic acid by GC- or LC-MS
Catalog No. 50-288-7197 Supplier Cayman Chemical Supplier No. 10742100 UG
An internal standard for the quantification of 5(Z),8(Z),11(Z)-eicosatrienoic acid by GC- or LC-MS