Learn More
Cayman Chemical 5 Z 8 ZTetradecadIenoIc Ac
SDP
Catalog No. 502919277
Encompass_Preferred
An internal standard for the quantification of 5(Z),8(Z)-tetradecadienoic acid by GC- or LC-MS
Catalog No. 50-291-9277 Supplier Cayman Chemical Supplier No. 39711250 UG
An internal standard for the quantification of 5(Z),8(Z)-tetradecadienoic acid by GC- or LC-MS