Learn More
Cayman Chemical 5 Z-DodecenoIc AcId 25mg
SDP
Catalog No. 502912802
Encompass_Preferred
A medium-chain unsaturated fatty acid; a precursor in the biosynthesis of the sex pheromone 5(Z)-dodecenal; levels are increased in culture medium from MCF-7 breast cancer cells compared to KMST-6 fibroblasts; plasma levels are increased in the plasma of patients with MCAD deficiency
Catalog No. 50-291-2802 Supplier Cayman Chemical Supplier No. 3591025 MG
A medium-chain unsaturated fatty acid; a precursor in the biosynthesis of the sex pheromone 5(Z)-dodecenal; levels are increased in culture medium from MCF-7 breast cancer cells compared to KMST-6 fibroblasts; plasma levels are increased in the plasma of patients with MCAD deficiency