Learn More
Biotium Mix-n-Stain™ Biotin Antibody Labeling Kit, 1x(50-100ug) labeling
Catalog No. NC1602167
Encompass_Preferred
Mix-n-Stain™ biotin antibody labeling kits are a revolutionary antibody labeling technology that allows you to label your antibody with biotin in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. Choose from kit sizes to label 5-20 ug, 20-50 ug, or 50-100 ug of your antibody.
Catalog No. NC1602167 Supplier Biotium Supplier No. 92244
Mix-n-Stain™ biotin antibody labeling kits are a revolutionary antibody labeling technology that allows you to label your antibody with biotin in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. Choose from kit sizes to label 5-20 ug, 20-50 ug, or 50-100 ug of your antibody.