Learn More
Ace Glass, Inc. 50/30 38mm ID 30cm chromatography column with 250mLreservoir, stopcock, and porosity B disc.
SDP
Catalog No. 50978601
Encompass_Preferred
38mm I.D., 30cm chromatography column with a 50/30 joint and a 250L integral reservoir. Column has a 2mm bore, 1:5 taper solid PTFE stopcock plug at bottom, and a porosity B (70-100 micron) fritted disc sealed in.
Catalog No. 50-978-601 Supplier Ace Glass, Inc. Supplier No. 590645
38mm I.D., 30cm chromatography column with a 50/30 joint and a 250L integral reservoir. Column has a 2mm bore, 1:5 taper solid PTFE stopcock plug at bottom, and a porosity B (70-100 micron) fritted disc sealed in.