Learn More
Ace Glass, Inc. 50/30 50mm ID 30cm chromatography column with 500mLreservoir and stopcock
SDP
Catalog No. 50972581
Encompass_Preferred
50mm I.D., 30cm chromatography column with a 50/30 joint and a 500L integral reservoir. Column has a 2mm bore, 1:5 taper solid PTFE stopcock plug at bottom.
Catalog No. 50-972-581 Supplier Ace Glass, Inc. Supplier No. 590618
50mm I.D., 30cm chromatography column with a 50/30 joint and a 500L integral reservoir. Column has a 2mm bore, 1:5 taper solid PTFE stopcock plug at bottom.