Learn More
Ace Glass, Inc. 50/30 50mm ID 30cm chromatography column with 500mLreservoir, stopcock, and porosity B disc.
SDP
Supplier:  Ace Glass, Inc. 590647
Encompass_Preferred
50mm I.D., 30cm chromatography column with a 50/30 joint and a 500L integral reservoir. Column has a 2mm bore, 1:5 taper solid PTFE stopcock plug at bottom, and a porosity B (70-100 micron) fritted disc sealed in.
Catalog No. 50-972-585