Learn More
Ace Glass, Inc. 50/30 ball joint, stainless steel, unthreaded
SDP
Catalog No. 50975302
Encompass
50/30, unthreaded, ball joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.
Catalog No. 50-975-302 Supplier Ace Glass, Inc. Supplier No. 765862
50/30, unthreaded, ball joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.